missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl phthalocyanine, 85%, pract.

Catalog No. AC422490250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422490250 25 g
AC422491000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422490250 Supplier Thermo Scientific Chemicals Supplier No. 422490250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13930-88-6 | C32H16N8OV | 579.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13930-88-6
Molecular Formula C32H16N8OV
Molecular Weight (g/mol) 579.46
InChI Key YRZZLAGRKZIJJI-UHFFFAOYSA-N
Synonym oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 %
PubChem CID 2735160
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 80 % min
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in xylene,2-ethylhexanoic acid,,2-ethylhexylamine and 6-methyl-2,4-heptanedione
Formula Weight 579.46
Percent Purity 85%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Vanadyl phthalocyanine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 237-700-2

RUO – Research Use Only