missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) Oxide, Extra Pure, MilliporeSigma™

Catalog No. M1008240250
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008240250 250 g Plastic bottle
M1008241000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1008240250 Supplier Technidata Supplier No. 1.00824.0250

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name [(dioxovanadio)oxy]dioxovanadium
SMILES O=[V](=O)O[V](=O)=O

Specifications

Physical Form Solid
Quantity 250 g
UN Number UN 2862
Formula Weight 181.88
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Appearance Yellow solid
Chemical Name or Material Vanadium(V) Oxide
Hazard Category H302 + H332: Harmful if swallowed or if inhaled
H335: May cause respiratory irritation.
H341: Suspected of causing genetic defects.
H361d: Suspected of damaging the unborn child.
H372: Causes damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-239-8