missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) oxide, 99+%

Catalog No. AC450162500
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450162500 250 g Plastic bottle
AC450160250 25 g Glass bottle
AC450160010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450162500 Supplier Thermo Scientific Chemicals Supplier No. 450162500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1314-62-1 | O5V2 | 181.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
SMILES O=[V](=O)O[V](=O)=O

Specifications

Boiling Point 1750.0°C
Melting Point 690.0°C
Quantity 250 g
Assay Percent Range 99% min
Linear Formula V2O5
Merck Index 15,10107
Solubility Information Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies, insoluble in alcohol
IUPAC Name dioxovanadiooxy(dioxo)vanadium
Formula Weight 181.88
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 3.35
Density 3.3500g/mL
Chemical Name or Material Vanadium(V) oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes damage to organs through prolonged or re
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
EINECSNumber 215-239-8

RUO – Research Use Only