missing translation for 'onlineSavingsMsg'
Learn More

Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis)

Catalog No. AA42190KM
Change view
Click to view available options
Quantity:
25 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42190KM 25 Pc.
AA42190KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42190KM Supplier Thermo Scientific Chemicals Supplier No. 042190KM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-62-2 | V | 50.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Quantity 25 Pc.
Odor Odorless
Weight ≈2.3g/slug
Form Slug
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Vanadium slug, 11mm (0.43 in.) dia. x 4mm (0.16 in.) thick
EINECSNumber 231-171-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only