missing translation for 'onlineSavingsMsg'
Learn More

Vanadium plate, 3.2mm (0.13in) thick, annealed, 99.5% (metals basis)

Catalog No. AA11099FI
Change view
Click to view available options
Quantity:
50 x 50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11099FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA11099FI Supplier Thermo Scientific Chemicals Supplier No. 011099FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-62-2 | V | 50.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈48.9g/50x50mm
Form Plate
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Vanadium plate, 3.2mm (0.13 in.) thick, Annealed
EINECSNumber 231-171-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only