missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide, 99% (metals basis)

Catalog No. p-4408207
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2295709 10 g
AA2295718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2295709 Supplier Thermo Scientific Chemicals Supplier No. 02295709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12036-21-4 | O2V | 82.94 g/mol

Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12036-21-4
Molecular Formula O2V
Molecular Weight (g/mol) 82.94
MDL Number MFCD00049702
InChI Key GRUMUEUJTSXQOI-UHFFFAOYSA-N
Synonym vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g
PubChem CID 82849
ChEBI CHEBI:30047
IUPAC Name dioxovanadium
SMILES O=[V]=O

Specifications

Melting Point 1967°C
Physical Form Powder
Quantity 10 g
Assay Percent Range 99% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 82.94
Density 4.339 g/mL
Chemical Name or Material Vanadium(IV) oxide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 234-841-1
TSCA Yes

RUO – Research Use Only