missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(III) fluoride, 98%
Description
Employed in oxygen-sensitive applications, such as metal production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ≥1406°C |
| Physical Form | Powder |
| Quantity | 2 g |
| UN Number | UN2923 |
| Merck Index | 14,9922 |
| Solubility Information | Practically insoluble in water, acetone, and glacial acetic acid |
| Formula Weight | 107.94 |
| Percent Purity | 98% |
| Sensitivity | Moisture sensitive |
| Density | 3.363 g/mL at 19°C |
| Show More |
Safety and Handling
| Hazard Category | H301-H314-H330-H335 |
| Hazard Statement | GHS H Statement H300-H330-H314-H318 Fatal if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| EINECSNumber | 233-169-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only