missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride, 99% (metals basis)

Catalog No. p-4408204
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1403909 10 g
AA1403918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1403909 Supplier Thermo Scientific Chemicals Supplier No. 01403909
Only null left

CAS: 7718-98-1 | Cl3V | 157.29 g/mol

Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7718-98-1
Molecular Formula Cl3V
Molecular Weight (g/mol) 157.29
MDL Number MFCD00011454
InChI Key HQYCOEXWFMFWLR-UHFFFAOYSA-K
PubChem CID 62647
IUPAC Name trichlorovanadium
SMILES [Cl-].[Cl-].[Cl-].[V+3]

Specifications

Quantity 10 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in tetrahydrofuran,alcohol,ether and hydrocarbons.
Formula Weight 157.3
Sensitivity Light and moisture sensitive
Density 3 g/mL
Chemical Name or Material Vanadium(III) chloride
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-744-6
RTECSNumber YW2800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only