missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Vanadium(III) chloride

Catalog No. 208272100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
208272100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 208272100G Supplier Sigma Aldrich Supplier No. 208272100G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vanadium trichloride acts as a reducing agent.Vanadium chloride may be used as a reduction agent to determine nitrates and nitrites in water samples.It may be used in redox flow batteries. Vanadium trichloride catalyses Pinacol coupling reactions of benzaldehyde in water. it also acts as a mild and efficient catalyst for the chemoselective deprotection of acetonides.

Specifications

CAS 7718-98-1
Quantity 100 g
Molecular Formula Cl3V
Linear Formula VCl3
MDL Number MFCD00011454
Synonym Vanadium trichloride
Molecular Weight (g/mol) 157.3
Percent Purity 97%
Density 3 g/mL (at 25°C)