missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) bromide, 99.5% (metals basis)

Catalog No. p-4408203
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8113504 2 g
AA8113509 10 g
AA8113518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8113504 Supplier Thermo Scientific Chemicals Supplier No. 08113504

CAS: 13470-26-3 | Br3V | 290.654 g/mol

Vanadium(III) bromide is used as a catalyst for C(sp3)-H fluorination reactions especially the benzylic fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13470-26-3
Molecular Formula Br3V
Molecular Weight (g/mol) 290.654
MDL Number MFCD00049695
InChI Key ZOYIPGHJSALYPY-UHFFFAOYSA-K
Synonym vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
PubChem CID 83509
IUPAC Name tribromovanadium
SMILES [V](Br)(Br)Br

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water and tetrahydrofuran.
Formula Weight 290.67
Sensitivity Moisture sensitive
Density 4.00^1^8 g/mL
Chemical Name or Material Vanadium(III) bromide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 236-736-6
RTECSNumber YW2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only