missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ValuGuard MP™ Lightweight Microporous Coveralls with Attached Hood and Boots
Economical coveralls are made of lightweight microporous fabric that provides high moisture vapor transmission rate (HMVT) to help workers stay cool and prevent heat stress.
$115.70 - $115.70
Specifications
| Product Type | General Purpose Coveralls |
|---|---|
| Gender | Unisex |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-184-045
|
International Enviroguard™
8119M |
Medium |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-046
|
International Enviroguard™
8119L |
Large |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-044
|
International Enviroguard™
8119XL |
X-Large |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-050
|
International Enviroguard™
81192XL |
2X-Large |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-049
|
International Enviroguard™
81193XL |
3X-Large |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-048
|
International Enviroguard™
81194XL |
4X-Large |
Case of 25 Each for $115.70
|
|
|||||
|
17-184-047
|
International Enviroguard™
81195XL |
5X-Large |
Case of 25 Each for $115.70
|
|
|||||
Description
- Fabric provides a high moisture vapor transmission rate (MVTR), helping you stay cooler to prevent heat stress
- Coveralls protect against particulates, light liquid splashes, and sprays in nonhazardous work environments
- Attached hood and boots offer full-body protection from head to toe
- Keep productivity high and risk level low when wearing comfortable ValuGuard MP Coveralls
- Secure, elastic wrists help keep sleeves in place to prevent skin exposure
- Standard zipper and set-in sleeves
- Environmentally friendly; can be recycled after use
Specifications
| General Purpose Coveralls | |
| Microporous Fabric | |
| Single-use | |
| Attached Boots | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Yes | |
| None |
| Unisex | |
| White | |
| Standard | |
| Elastic | |
| Attached, Elastic Face | |
| Serged | |
| Tears, Liquid, Particle | |
| 8119 |