Learn More
DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class B, with Snap Cap
Catalog No. S00139
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S00139 1000 mL
S00136 100 mL
S00137 250 mL
S00135 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S00139 Supplier DWK Life Sciences Supplier No. 580101000
Calibrated To Contain.
  • Kimax™ KG-33 glass
  • Graduation ring blasted on neck, tooled for included polyethylene snap cap
  • Enlarged top of the cap protects neck if flask is tipped over
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class B, unserialized requirements.
Specifications
Calibration Calibrated To Contain
Closure Material Polyethylene
Capacity (Metric) 1000 mL
Material Glass
Closure Size 6