Learn More
DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
Calibrated To Contain.
Supplier:  DWK Life Sciences 58014200
 View more versions of this product
Catalog No. S00124
Only null left
Explore available promotions
Description
Description
  • Kimax™ KG-33 glass Single graduation ring blasted on the neck
  • With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Specifications
Calibrated To Contain
200 mL
16
Glass
Glass
Product Suggestions
Product Suggestions