Learn More
DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
Calibrated To Contain.
Supplier: DWK Life Sciences 5801450
Description
- Kimax™ KG-33 glass Single graduation ring blasted on the neck
- With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
| Calibrated To Contain | |
| 50 mL | |
| 9 |
| Glass | |
| Glass |