missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Value Convertible Refrigerator/Freezer

Catalog No. fbv20rpsa
Change view
Click to view available options
Capacity (Metric) Refrigerator:
594 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Refrigerator
FBV20RPSA 594 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FBV20RPSA Supplier Fisherbrand™ Supplier No. FBV20RPSA
Only null left

Flexible and economical laboratory storage.

Fisherbrand™ Value Convertible Refrigerator/Freezers offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.

Note: Comes configured as a refrigerator

Specifications

Type General Purpose Refrigerator (Convertible to Freezer)
Capacity (English) Refrigerator 21 cu. ft.
Capacity (Metric) Refrigerator 594 L
Temperature Range (Freezer) -12°C to -24°C
Temperature Range (Refrigerator) 1°C to 12°C
Temperature (Setpoint) Adjustable
Door Style Hinged, Open Left to Right, Non-reversible
No. of Doors 1 (1 compartment)
Shelves 3 Adjustable
Door Shelves 4 fixed shelves
Compressor Bottom-mounted
Defrost Automatic
Operating Modes 2 - Refrigerator or Freezer
Refrigerant R600a
Exterior Finish White-enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Casters 2 adjustable leveling feet
Color White
Control Type Electronic control
For Use With (Application) Indoor general storage of non-critical samples and reagents
Lighting Interior
Min. Clearance 4 in. on sides, 4 in. at back, 11 in. on top
Typical Lifetime 10 years
Warranty 13 months parts and labor
Dimensions (DxWxH) Exterior 73.5 x 83.2 x 195 cm (28.94 x 32.76 x 76.77 in.)
Dimensions (D x W x H) Interior 57.15 x 67.3 x 175.26 cm (22.5 x 26.5 x 69 in.)
Amperage 2.2 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V/60 Hz
Frequency 50 Hz
Shipping Weight (English) 244 lb.
Shipping Weight (Metric) 110.6 kg
Show More Show Less