missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Valsartan | |
| 117°C | |
| C24H29N5O3 | |
| 5 g | |
| ACWBQPMHZXGDFX-QFIPXVFZSA-N | |
| (2S)-3-methyl-2-[pentanoyl-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid | |
| 60846 | |
| 435.53 | |
| Crystalline Powder |
| 137862-53-4 | |
| White | |
| MFCD00865840 | |
| N-Valeryl-N-[2′-(1H-tetrazol-5-yl)biphenyl-4-ylmethyl]-L-valine | |
| CCCCC(=O)N(CC1=CC=C(C=C1)C2=CC=CC=C2C3=NNN=N3)C(C(C)C)C(=O)O | |
| 435.528 | |
| CHEBI:9927 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 137862-53-4 | |
| 435.528 | |
| ACWBQPMHZXGDFX-QFIPXVFZSA-N | |
| CHEBI:9927 | |
| CCCCC(=O)N(CC1=CC=C(C=C1)C2=CC=CC=C2C3=NNN=N3)C(C(C)C)C(=O)O |
| C24H29N5O3 | |
| MFCD00865840 | |
| 60846 | |
| (2S)-3-methyl-2-[pentanoyl-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid |
Safety and Handling
RTECSNumber : YV9455000
TSCA : No