missing translation for 'onlineSavingsMsg'
Learn More

Valsartan 98.0+%, TCI America™

Catalog No. V01121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01121G 1 g
V01125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01121G Supplier TCI America Supplier No. V01121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137862-53-4
Molecular Formula C24H29N5O3
Molecular Weight (g/mol) 435.528
MDL Number MFCD00865840
InChI Key ACWBQPMHZXGDFX-QFIPXVFZSA-N
PubChem CID 60846
ChEBI CHEBI:9927
IUPAC Name (2S)-3-methyl-2-[pentanoyl-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid
SMILES CCCCC(=O)N(CC1=CC=C(C=C1)C2=CC=CC=C2C3=NNN=N3)C(C(C)C)C(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Synonym N-Valeryl-N-[2′C-(1H-tetrazol-5-yl)biphenyl-4-ylmethyl]-L-valine
Formula Weight 435.53
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Valsartan
RTECSNumber YV9455000
TSCA No