missing translation for 'onlineSavingsMsg'
Learn More

Valproic Acid Impurity K, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023025
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023025 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023025 Supplier Merck Emd Millipore Supplier No. PHR2282100MG

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 5343-52-2
Molecular Formula C8H16O2
Linear Formula C8H16O2
MDL Number MFCD01675509
Quantity 100 mg
Synonym (2 RS)-2-Ethyl-2-methylpentanoic acid; 2-Ethyl-2-methylvaleric acid; NSC 466
Formula Weight 144.21
Grade Certified Reference Material