missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valnemulin Hydrochloride 98.0+%, TCI America™
$301.98 - $1056.92
Chemical Identifiers
| CAS | 133868-46-9 |
|---|---|
| Molecular Formula | C31H53ClN2O5S |
| Molecular Weight (g/mol) | 601.284 |
| MDL Number | MFCD08458958 |
| InChI Key | MFBPRQKHDIVLOJ-AFFLPQGKSA-N |
| PubChem CID | 60195218 |
| SMILES | CC1CCC23CCC(=O)C2C1(C(CC(C(C3C)O)(C)C=C)OC(=O)CSC(C)(C)CNC(=O)C(C(C)C)N)C.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V014450MG
|
TCI America
V014450MG |
50 mg |
Each for $301.98
|
|
|||||
|
V0144250MG
|
TCI America
V0144250MG |
250 mg |
Each for $1,056.92
|
|
|||||
Chemical Identifiers
| 133868-46-9 | |
| 601.284 | |
| MFBPRQKHDIVLOJ-AFFLPQGKSA-N | |
| CC1CCC23CCC(=O)C2C1(C(CC(C(C3C)O)(C)C=C)OC(=O)CSC(C)(C)CNC(=O)C(C(C)C)N)C.Cl |
| C31H53ClN2O5S | |
| MFCD08458958 | |
| 60195218 |
Specifications
| 133868-46-9 | |
| C31H53ClN2O5S | |
| 50 mg | |
| CC1CCC23CCC(=O)C2C1(C(CC(C(C3C)O)(C)C=C)OC(=O)CSC(C)(C)CNC(=O)C(C(C)C)N)C.Cl | |
| 60195218 | |
| ≥98.0% (HPLC) | |
| Valnemulin Hydrochloride |
| White | |
| MFCD08458958 | |
| MFBPRQKHDIVLOJ-AFFLPQGKSA-N | |
| 601.284 | |
| 601.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)