missing translation for 'onlineSavingsMsg'
Learn More

Valnemulin Hydrochloride 98.0+%, TCI America™

Catalog No. V014450MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V014450MG 50 mg
V0144250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V014450MG Supplier TCI America Supplier No. V014450MG

Chemical Identifiers

CAS 133868-46-9
Molecular Formula C31H53ClN2O5S
Molecular Weight (g/mol) 601.284
MDL Number MFCD08458958
InChI Key MFBPRQKHDIVLOJ-AFFLPQGKSA-N
PubChem CID 60195218
SMILES CC1CCC23CCC(=O)C2C1(C(CC(C(C3C)O)(C)C=C)OC(=O)CSC(C)(C)CNC(=O)C(C(C)C)N)C.Cl

Specifications

Color White
Quantity 50 mg
Formula Weight 601.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Valnemulin Hydrochloride
TSCA No
Recommended Storage Freezer (-20 °C)