missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valnemulin hydrochloride

Catalog No. AC462832500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462832500 250 mg
AC462830010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462832500 Supplier Thermo Scientific Chemicals Supplier No. 462832500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133868-46-9
Molecular Formula C31H53ClN2O5S
Molecular Weight (g/mol) 601.28
InChI Key MFBPRQKHDIVLOJ-UHFFFAOYNA-N
IUPAC Name 4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[1-(2-amino-3-methylbutanamido)-2-methylpropan-2-yl]sulfanyl}acetate hydrochloride
SMILES Cl.CC(C)C(N)C(=O)NCC(C)(C)SCC(=O)OC1CC(C)(C=C)C(O)C(C)C23CCC(=O)C2C1(C)C(C)CC3

Specifications

Quantity 250 mg
Formula Weight 601.29
Chemical Name or Material Valnemulin hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

May cause respiratory irritation.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear eye protection/face protection.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only