missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valinomycin, ≥93%, MP Biomedicals™
Specifications
Specifications
| Type | Valinomycin |
| Physical Form | Crystalline Powder |
| Color | White |
| Quantity | 10 mg |
| Assay Percent Range | ≥93% |
| Formula Weight | 1111.4 |
| Grade | Not Available |
| Chemical Name or Material | Valinomycin |