missing translation for 'onlineSavingsMsg'
Learn More

Valinomycin, ≥93%, MP Biomedicals™

Catalog No. ICN10501005
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10501005 5 mg
ICN10501010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10501005 Supplier MP Biomedicals Inc Supplier No. 10501005

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.34
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
PubChem CID 131668508
IUPAC Name (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

Specifications

Type Valinomycin
Physical Form Crystalline Powder
Color White
Quantity 5 mg
Assay Percent Range ≥93%
Formula Weight 1111.4
Chemical Name or Material Valinomycin