missing translation for 'onlineSavingsMsg'
Learn More

Valinomycin, 90+%

Catalog No. AAJ62312MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62312MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62312MC Supplier Thermo Scientific Chemicals Supplier No. J62312MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2001-95-8 | C54H90N6O18 | 1111.34 g/mol

Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

Solubility
Soluble in DMSO at 10mg/ml. Insoluble in w

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.34
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
Synonym NSC 122023
PubChem CID 131668508
IUPAC Name (3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

Specifications

Type Valinomycin
Physical Form Crystalline Powder
Color White
Quantity 100 mg
UN Number UN2811
Beilstein 78657
Merck Index 14,9910
Solubility Information Soluble in DMSO at 10mg/ml; Insoluble in water
Formula Weight 1111.34
Percent Purity ≥90%
Chemical Name or Material Valinomycin
Show More Show Less
Hazard Category H300+H310
Hazard Statement GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement P262-P264b-P270-P280-P301+P310-P302+P350-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-896-6
RTECSNumber YV9468000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only