missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valeronitrile 98.0+%, TCI America™
$51.98 - $381.18
Chemical Identifiers
| CAS | 110-59-8 |
|---|---|
| Molecular Formula | C5H9N |
| Molecular Weight (g/mol) | 83.134 |
| MDL Number | MFCD00001974 |
| InChI Key | RFFFKMOABOFIDF-UHFFFAOYSA-N |
| Synonym | valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile |
| PubChem CID | 8061 |
| IUPAC Name | pentanenitrile |
| SMILES | CCCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V000825ML
|
TCI America
V000825ML |
25 mL |
Each for $51.98
|
|
|||||
|
V0008500ML
|
TCI America
V0008500ML |
500 mL |
Each for $381.18
|
|
|||||
Chemical Identifiers
| 110-59-8 | |
| 83.134 | |
| RFFFKMOABOFIDF-UHFFFAOYSA-N | |
| 8061 | |
| CCCCC#N |
| C5H9N | |
| MFCD00001974 | |
| valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
| pentanenitrile |
Specifications
| 110-59-8 | |
| Yellow | |
| C5H9N | |
| 25 mL | |
| valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
| CCCCC#N | |
| 83.134 | |
| 83.13 | |
| Liquid |
| -96°C | |
| 140°C | |
| MFCD00001974 | |
| 3273 | |
| RFFFKMOABOFIDF-UHFFFAOYSA-N | |
| pentanenitrile | |
| 8061 | |
| ≥98.0% (GC) | |
| Valeronitrile |
Safety and Handling
EINECSNumber : (2)-3805
RTECSNumber : YV8195000
TSCA : Yes