missing translation for 'onlineSavingsMsg'
Learn More

Valeronitrile 98.0+%, TCI America™

Catalog No. V0008500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0008500ML 500 mL
V000825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0008500ML Supplier TCI America Supplier No. V0008500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-59-8
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001974
InChI Key RFFFKMOABOFIDF-UHFFFAOYSA-N
Synonym valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile
PubChem CID 8061
IUPAC Name pentanenitrile
SMILES CCCCC#N

Specifications

Melting Point -96°C
Color Yellow
Boiling Point 140°C
Quantity 500 mL
UN Number 3273
Formula Weight 83.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Valeronitrile
EINECSNumber (2)-3805
RTECSNumber YV8195000
TSCA Yes