Learn More
Valeronitrile, 99%
CAS: 110-59-8 | C5H9N | 83.134 g/mol
$51.19 - $503.11
Chemical Identifiers
| CAS | 110-59-8 |
|---|---|
| Molecular Formula | C5H9N |
| Molecular Weight (g/mol) | 83.134 |
| MDL Number | MFCD00001974 |
| InChI Key | RFFFKMOABOFIDF-UHFFFAOYSA-N |
| Synonym | valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile |
| PubChem CID | 8061 |
| IUPAC Name | pentanenitrile |
| SMILES | CCCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA10306AC
|
Thermo Scientific Chemicals
A10306AC |
25 mL |
Each for $51.19
|
|
|||||
|
AAA10306AE
|
Thermo Scientific Chemicals
A10306AE |
100 mL |
Each for $142.47
|
|
|||||
|
AAA10306AP
|
Thermo Scientific Chemicals
A10306AP |
500 mL |
Each for $503.11
|
|
|||||
Description
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsValeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.
Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 110-59-8 | |
| 83.134 | |
| RFFFKMOABOFIDF-UHFFFAOYSA-N | |
| 8061 | |
| CCCCC#N |
| C5H9N | |
| MFCD00001974 | |
| valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
| pentanenitrile |
Specifications
| 110-59-8 | |
| 0.797 | |
| 38°C (100°F) | |
| C5H9N | |
| MFCD00001974 | |
| UN1992 | |
| 14,9905 | |
| Soluble in water at 20°C 10g/L. Soluble in alcohol. | |
| CCCCC#N | |
| 83.134 | |
| 83.13 | |
| Valeronitrile |
| -96°C | |
| 139°C to 141°C | |
| Odorless | |
| 1.397 | |
| 25 mL | |
| 1736706 | |
| valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
| RFFFKMOABOFIDF-UHFFFAOYSA-N | |
| pentanenitrile | |
| 8061 | |
| 99% |
Safety and Handling
GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 203-781-8
RTECSNumber : YV8195000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only