missing translation for 'onlineSavingsMsg'
Learn More

Valeronitrile, 99%

Catalog No. AAA10306AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10306AC 25 mL
AAA10306AE 100 mL
AAA10306AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10306AC Supplier Thermo Scientific Chemicals Supplier No. A10306AC

CAS: 110-59-8 | C5H9N | 83.134 g/mol

Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 110-59-8
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001974
InChI Key RFFFKMOABOFIDF-UHFFFAOYSA-N
Synonym valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile
PubChem CID 8061
IUPAC Name pentanenitrile
SMILES CCCCC#N

Specifications

Melting Point -96°C
Density 0.797
Boiling Point 139°C to 141°C
Flash Point 38°C (100°F)
Odor Odorless
Refractive Index 1.397
Quantity 25 mL
UN Number UN1992
Beilstein 1736706
Merck Index 14,9905
Solubility Information Soluble in water at 20°C 10g/L. Soluble in alcohol.
Formula Weight 83.13
Percent Purity 99%
Chemical Name or Material Valeronitrile
Show More Show Less
Hazard Category H226-H302
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-781-8
RTECSNumber YV8195000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only