missing translation for 'onlineSavingsMsg'
Learn More

Valeronitrile, 98%

Catalog No. AC167941000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167941000 100 mL Glass bottle
AC167945000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167941000 Supplier Thermo Scientific Chemicals Supplier No. 167941000

CAS: 110-59-8 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-59-8
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001974
InChI Key RFFFKMOABOFIDF-UHFFFAOYSA-N
Synonym valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile
PubChem CID 8061
IUPAC Name pentanenitrile
SMILES CCCCC#N

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.7900g/mL
Boiling Point 139.0°C to 141.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3960 to 1.3980
Linear Formula CH3(CH2)3CN
Quantity 100 mL
Beilstein 02, 301
Specific Gravity 0.79
Merck Index 15, 10091
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valeronitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-781-8

RUO – Research Use Only