missing translation for 'onlineSavingsMsg'
Learn More

Valeric acid, 99%

Catalog No. AAA16238AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16238AP 500 mL
AAA16238AE 100 mL
AAA162380F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA16238AP Supplier Thermo Scientific Chemicals Supplier No. A16238AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-52-4 | C5H10O2 | 102.133 g/mol

Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O

Specifications

Melting Point -34°C
Boiling Point 184°C to 186°C
Density 0.938
Quantity 500 mL
Flash Point 88°C (190°F)
Odor Unpleasant
Refractive Index 1.408
Linear Formula CH3(CH2)3CO2H
UN Number UN3265
Beilstein 969454
Merck Index 14,9904
Solubility Information It is soluble in water.
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material Valeric acid
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-677-2
RTECSNumber YV6100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only