missing translation for 'onlineSavingsMsg'
Learn More

Valeric Acid, 99%, Spectrum™ Chemical

Catalog No. 18614635
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614635 250 mL Amber Glass Bottle
18614634 1 L Amber Glass Bottle
18614636 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614635 Supplier Spectrum Chemical Mfg Cor Supplier No. VA110250ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 109-52-4

Specifications

CAS Max % 1
Percent Purity 99%
Grade Reagent
Quantity 250 mL
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.