missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valeric Acid, 99%

CAS: 109-52-4 | C5H10O2 | 102.133 g/mol

$112.23 - $212.70

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149570050
SDS View Documents
Thermo Scientific Chemicals
149570050
5 mL Glass bottle
Each for $112.23
Only null left
 
AC149571000
SDS View Documents
Thermo Scientific Chemicals
149571000
100 mL Glass bottle
Each for $127.74
Only null left
 
AC149570010
SDS View Documents
Thermo Scientific Chemicals
149570010
1 L Glass bottle
Each for $163.80
Only null left
 
AC149570025
SDS View Documents
Thermo Scientific Chemicals
149570025
2.5 L Glass bottle
Each for $212.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-52-4
102.133
NQPDZGIKBAWPEJ-UHFFFAOYSA-N
7991
pentanoic acid
C5H10O2
MFCD00004413
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
CHEBI:17418
CCCCC(=O)O

Specifications

-20.0°C to +18.0°C
185.0°C
99%
CH3(CH2)3CO2H
02, 299
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
NQPDZGIKBAWPEJ-UHFFFAOYSA-N
pentanoic acid
7991
102.13
0.9400g/mL
86°C
Glass bottle
1.4070 to 1.4100
Valeric acid
109-52-4
Colorless to Yellow
C5H10O2
MFCD00004413
15, 10090
Solubility in water: 40g/L (20°C). Other solubilities: soluble in alcohol and ether
CCCCC(=O)O
102.133
CHEBI:17418
99%
5 mL
Authentic
Liquid
0.94
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

SDS
Documents

Documents

RUO – Research Use Only