missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valeric Acid, 99%

Catalog No. ac149570050
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149570050 5 mL Glass bottle
AC149571000 100 mL Glass bottle
AC149570010 1 L Glass bottle
AC149570025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC149570050 Supplier Thermo Scientific Chemicals Supplier No. 149570050
Only null left

CAS: 109-52-4 | C5H10O2 | 102.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O

Specifications

Melting Point -20.0°C to +18.0°C
Boiling Point 185.0°C
Color Colorless to Yellow
Assay Percent Range 99%
Linear Formula CH3(CH2)3CO2H
Beilstein 02, 299
Merck Index 15, 10090
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 102.13
Percent Purity 99%
Density 0.9400g/mL
Quantity 5 mL
Flash Point 86°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4070 to 1.4100
Specific Gravity 0.94
Chemical Name or Material Valeric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

RUO – Research Use Only