missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Valeramide | |
| 105°C | |
| C5H11NO | |
| 250 g | |
| IPWFJLQDVFKJDU-UHFFFAOYSA-N | |
| pentanamide | |
| 12298 | |
| 101.15 | |
| Crystalline Powder |
| 626-97-1 | |
| White | |
| MFCD00041895 | |
| valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard | |
| CCCCC(=O)N | |
| 101.149 | |
| CHEBI:16459 | |
| ≥98.0% (N) |
Chemical Identifiers
| 626-97-1 | |
| 101.149 | |
| IPWFJLQDVFKJDU-UHFFFAOYSA-N | |
| 12298 | |
| pentanamide |
| C5H11NO | |
| MFCD00041895 | |
| valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard | |
| CHEBI:16459 | |
| CCCCC(=O)N |
Safety and Handling
TSCA : Yes