missing translation for 'onlineSavingsMsg'
Learn More

Valeramide, 97%

Catalog No. AC422440250
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422440250 25 g Glass bottle
AC422440025 2.5 g Glass bottle
AC422440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC422440250 Supplier Thermo Scientific Chemicals Supplier No. 422440250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-97-1 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626-97-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00041895
InChI Key IPWFJLQDVFKJDU-UHFFFAOYSA-N
Synonym valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard
PubChem CID 12298
ChEBI CHEBI:16459
IUPAC Name pentanamide
SMILES CCCCC(=O)N

Specifications

Melting Point 101°C to 106°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CONH2
Quantity 25 g
Formula Weight 101.15
Percent Purity 97%
Physical Form Shiny Powder or Flakes
Chemical Name or Material Valeramide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-974-0
TSCA TSCA

RUO – Research Use Only