missing translation for 'onlineSavingsMsg'
Learn More

Valeramide, 97%

Catalog No. p-7046498
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1460106 5 g
AAL1460114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1460106 Supplier Thermo Scientific Chemicals Supplier No. L1460106
Only null left

CAS: 626-97-1 | C5H11NO | 101.149 g/mol

Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
Soluble in water (almost transparency).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 626-97-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00041895
InChI Key IPWFJLQDVFKJDU-UHFFFAOYSA-N
Synonym valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard
PubChem CID 12298
ChEBI CHEBI:16459
IUPAC Name pentanamide
SMILES CCCCC(=O)N

Specifications

Melting Point 102°C to 104°C
Quantity 5 g
Beilstein 1740792
Solubility Information Soluble in water (almost transparency).
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material Valeramide
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-974-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only