Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Catalog No. 03170438
Change view
Click to view available options
Description:
Ferrule
Male Nut
Quantity:
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
03170438 Ferrule 10 Pack
03170437 Male Nut 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03170438 Supplier Thermo Scientific™ Supplier No. U401X
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
  • Designed for 10-32 port configurations
  • Burr and contaminant free
Specifications
Description Ferrule
Material Stainless Steel
For Use With (Equipment) 1/16” tubing
Size (English) 0.625 in.
Type Universal Ferrule
Quantity 10 Pack
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.