missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valacyclovir Hydrochloride Hydrate 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 360.80 |
| Percent Purity | ≥97.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Valacyclovir Hydrochloride Hydrate |
Safety and Handling
| TSCA | No |