missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ [Val5] Angiotensin II, Human
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 1032.15 |
| Product Type | [Val5] Angiotensin II, human |
| Quantity | 5 mg |
| Regulatory Status | RUO |
| Sequence | Asp-Arg-Val-Tyr-Val-His-Pro-Phe |
For Research Use Only