missing translation for 'onlineSavingsMsg'
Learn More

VAI Steri-Oil Mineral Oil Lubricant

Catalog No. 19546939
Change view
Click to view available options
Volume (Metric):
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19546939 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19546939 Supplier VAI Supplier No. SO200A1Z

missing translation for 'validMainframeMsg'

Steri-Oil Mineral Oil Lubricant is a sterile, USP grade, penetrating mineral oil lubricant. STERI-OIL 200 has been developed for use as a sterile lubricant in aseptic manufacturing areas and cleanrooms to reduce items from sticking, penetrate and lubricate mechanisms, and for moisture displacement.

  • Specifically formulated as a sterile cleanroom formula
  • Available in an 1 oz. bottle with a dropper tip for precise application
  • Bottle design prevents over use and over application
  • Heavy consistency ensures excellent lubrication and penetration
  • Allows for trouble free processing during manufacturing
  • 100% mineral oil

Specifications

Product Type Mineral Oil Lubricant
For Use With (Application) Aseptic manufacturing, prevents metal/metal contact
Packaging Double Bagged
Quantity 250/Cs.
Volume (English) 1 oz.
Volume (Metric) 30 mL