missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAI Process2Clean 5 Emulsifying Detergent/Additive
Process2Clean 5 Emulsifying Detergent/Additive is a highly built neutral pH cleaning agent that can be used as a stand-alone detergent or as an additive to improve the cleaning performance of all VAI cleaning agents.
$861.53 - $861.53
Specifications
| Appearance | Pale Yellow |
|---|---|
| For Use With (Application) | For high/low energy clean-in-place products, recirculation/high pressure cleaning |
| Catalog Number | Mfr. No. | Volume (English) | Sterility | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (English) | Sterility | Price | Quantity | ||||
|
19-546-898
|
VAI
PC51G01 |
1 gal. | Non-sterile |
Case of 4 Each for $861.53
|
|
||||
Description
- Characteristic: Neutral pH
- Clarity: Clear
- Appearance: Yellow
- pH (1% solution) : 7.5 â 9.0
- Conductivity (1% solution): 0.05 â 0.20 mS
Specifications
| Pale Yellow | |
| For high/low energy clean-in-place products, recirculation/high pressure cleaning |