Learn More
Chemglass Inc Vacuum Trap, Removable Arms, Complete
Catalog No. CG452701
Encompass
Vacuum Trap, Removable Arms, Complete. Vacuum trap features removable arms to permit the trap to be easily disconnected for cleaning. The arms are made to permit clamping directly to the lattice and accept 5/8in I.D. vacuum hose. Trap and arms have 35/25 spherical joints for added flexibility while providing a leak-free seal. Supplied complete with one each of arm style A (180), one each of arm style B (90) and one each of arm style C (straight), and two CG-150-06 #35 stainless steel pinchclamps. The trap body is 2in (50mm) O.D. x 360mm long.
Catalog No. CG452701 Supplier Chemglass Inc Supplier No. CG452701
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum Trap, Removable Arms, Complete. Vacuum trap features removable arms to permit the trap to be easily disconnected for cleaning. The arms are made to permit clamping directly to the lattice and accept 5/8in I.D. vacuum hose. Trap and arms have 35/25 spherical joints for added flexibility while providing a leak-free seal. Supplied complete with one each of arm style A (180), one each of arm style B (90) and one each of arm style C (straight), and two CG-150-06 #35 stainless steel pinchclamps. The trap body is 2in (50mm) O.D. x 360mm long.