Learn More
Chemglass Inc Vacuum Trap, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt.
Catalog No. CG451410
Encompass
Vacuum Trap, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.
Catalog No. CG451410 Supplier Chemglass Inc Supplier No. CG451410
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum Trap, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.