Learn More
Chemglass Inc Vacuum Trap, Solvent Vapor, Plastic Coated, Complete
Catalog No. CG451901
Encompass
Vacuum Trap, Solvent Vapor, Plastic Coated, Complete. Trap is designed for large volume solvent vapor removal during rotary evaporation. Trap is placed between the vacuum source and the rotary evaporator to prevent solvent vapors from entering the vacuum pump. PLASTIC COATED to contain solvent in case of breakage and supplied complete with a UHMW lid with handle.
Catalog No. CG451901 Supplier Chemglass Inc Supplier No. CG451901
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum Trap, Solvent Vapor, Plastic Coated, Complete. Trap is designed for large volume solvent vapor removal during rotary evaporation. Trap is placed between the vacuum source and the rotary evaporator to prevent solvent vapors from entering the vacuum pump. PLASTIC COATED to contain solvent in case of breakage and supplied complete with a UHMW lid with handle.