Learn More
BINDER™ Vacuum Pumps VAP with Pump Module
Description
- Choice of chemistry diaphragm pumps that are optimally matched to the vacuum ovens.
- Scoop is resistant to a variety of chemicals, especially acids
- Final vacuum can be regulated via the vacuum oven
- Pump module lifts the vacuum oven to the optimum working height, significantly reduces the noise of the vacuum pump, and protects the pump and glass material from mechanical damage.
- Pump in the module can be switched on and off using a switchable socket via the control of the vacuum furnace.
Specifications
Specifications
| For Use With (Equipment) | Model VD 23 |
| Material | Steel |
| Type | VAP 1 |
| Includes | Pump module |
| Electrical Requirements | 50/60 Hz |
| Voltage | 120 V |