missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Vacuum Pumps VAP

Choice of chemistry diaphragm pumps that are optimally matched to the vacuum ovens.

Supplier:  BINDER™ 50130223

 View more versions of this product

Catalog No. 08111934

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Scoop is resistant to a variety of chemicals, especially acids.
  • Final vacuum can be regulated via the vacuum oven.
  • Chemistry diaphragm pumps VAP, ATEX compliant acc.: II 3G IIC T3 X (only for the inner chamber and wetted parts)
Specifications

Chemical Identifiers

BINDER Vacuum Ovens
Steel
120 V
50/60 Hz

Specifications

Nominal pumping speed: 2 m³/h
Steel
Final pressure: 7 Mbar
120 V
BINDER Vacuum Ovens
Chemistry Diaphragm Pumps VAP 1
Pump Speed: 2 m³/h
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions