Learn More
BUCHI Vacuum Pump V-600 for Rotavapor R-220 Pro Industrial Evaporator for multiple benchtop Rotavapors
Vacuum Pump V-600 is a chemically resistant 3-stage diaphragm pump with silent and economical operation that makes it suited to work with the R-220 Pro or multiple benchtop Rotavapors
Supplier:  BUCHI 11V600200
 View more versions of this product
Catalog No. 05406013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Chemically resistant 3-stage diaphragm pump
  • Quiet and economical operation
  • Optimally suited to work with the R-220 Pro, multiple Rotavapors or SyncorePlus Parallel Evaporation
  • 1.5 mbar final vacuum
  • 3.1 m3/h vacuum capacity
Specifications
Chemical Identifiers
1.5 mbar
40 to 64 dBA
Vacuum System
V-600
50/60 Hz
IP20
Specifications
1.5 mbar
V-600 Vacuum Pump, I-300 Interface, Woulff Bottle
V-600
3.1 m³/h
Vacuum System
Rotary evaporation
I-300 Interface
332 x 330 x 291 mm
40 to 64 dBA
50/60 Hz
IP20
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CE, CSA, CUS