Learn More
BINDER™ Vacuum Module with Vacuum Pump VP 2
Catalog No. 11103200
Change view
Click to view available options
For Use With:
BINDER units VD023UL-120V
BINDER units VD056UL-120V
BINDER units VD115UL-120V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11103200 BINDER units VD056UL-120V
11103193 BINDER units VD115UL-120V
11103198 BINDER units VD023UL-120V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11103200 Supplier BINDER™ Supplier No. 80122623
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum module with vacuum pump VP 2 - 120 V / 50-60 Hz option model, Pump chamber for installation of vacuum pumps, with switchable power socket.
  • Temperature range:
  • +9°C above ambient temperature to +110 °C
  • Controller with digital display of pressure and temperature
  • Program-controlled drying monitoring with automatic ventilation at end of process
  • Internal data logger, measured values can be read out in open format via USB
  • 1 aluminum expansion rack, can be custom-positioned
  • Inert gas connection
  • Universal access port DN 16 - for the introduction of external measuring devices
  • Shatterproof, spring-mounted safety glass panel
  • Large viewing window with option of interior lighting
Specifications
Amperage 3 to 5.7
Electrical Requirements 50/60 Hz
Dimensions (L x W x H) 581 x 743 x 942 mm
Temperature (Metric) Max 110°C
For Use With BINDER units VD056UL-120V
Voltage 120 V