Learn More
GVS™ Vacuum Manifold Stainless Steel
Vacuum manifold used for filtration and analysis of liquid samples.
Supplier: GVS™ 10498763
Description
Description
- Stainless steel filter funnel three-place or six-place manifold, for use with microbiology monitors, analytical funnels and general filtration
Features and Benefits
- Durable Stainless Steel construction for easy cleanup and compatibility with many chemicals.
- Works with all GVS BioFunnel manifolds and analytical funnels.
- Resistant to sample
- Portable
Applications:
- Used for portable water analysis, waste water analysis, dairy product analysis, beverages analysis, general filtration of liquid samples.
- Allows performance of tests simultaneously.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
| Manifold | |
| Stainless Steel | |
| 3-position | |
| 1 |