Learn More
Chemglass Inc Vacuum Gauge, Digital, 110/240 Volt, Complete
Supplier: Chemglass Inc AF030301
Vacuum Gauge, Digital, 110/240 Volt, Complete. This unit measures vacuum in Torr and uses a rugged thermocouple vacuum gauge tube. The gauge controller is housed in a laboratory grade case constructed of vinyl clad metal on an extruded aluminum base ensuring maximum durability. The gauge is highly resistant to shock because it has no moving parts, thus requiring infrequent calibration. The gauge controller has a large bright LED display which can be easily read from across the room or in dimly lit locations. This gauge measures from 1 micron (1x10-3 Torr) all the way up to 760,000 Microns (760 Torr)or Atmosphere. The wide range of this vacuum instrument enables users to easily troubleshoot their system Gauge tube is threaded with 1/8 male NPT for use with AF-0306 adapter.